Assay Details
Gene Symbol: NR3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013978839.2 | XP_013834293.1 | 3117 | 63 | ||
XM_013978840.2 | XP_013834294.1 | 3838 | 63 | |||
XM_013978843.2 | XP_013834297.1 | 3178 | 63 | |||
XM_021100657.1 | XP_020956316.1 | 3190 | 63 | |||
GenBank mRNA | AK390253.1 | - | 6 | 899 | 63 | |
U88893.1 | - | 3 | 345 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
