Assay Details
Gene Symbol: SOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190422.1 | NP_001177351.1 | 2 - 3 | 240 | 113 | |
GenBank mRNA | AF396674.1 | - | 2 - 3 | 155 | 113 | |
AK236139.1 | - | 2 - 3 | 113 | |||
AK240282.1 | - | 2 - 3 | 244 | 113 | ||
AK346331.1 | - | 2 - 3 | 113 | |||
AK351289.1 | - | 2 - 3 | 248 | 113 | ||
AK392550.1 | - | 2 - 3 | 113 | |||
AK392718.1 | - | 2 - 3 | 242 | 113 | ||
AK400130.1 | - | 2 - 3 | 268 | 113 | ||
GQ913661.1 | - | 2 - 3 | 214 | 113 | ||
GU944822.1 | - | 2 - 3 | 240 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
