Assay Details
Gene Symbol: ADIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007193.1 | NP_001007194.1 | 6 | 948 | 73 | |
XM_005668010.3 | XP_005668067.2 | 1185 | 73 | |||
GenBank mRNA | AB527058.1 | - | 6 | 935 | 73 | |
AK232692.1 | - | 7 | 1119 | 73 | ||
AK235133.1 | - | 7 | 1125 | 73 | ||
AK239374.1 | - | 3 | 457 | 73 | ||
AY452710.1 | - | 6 | 858 | 73 | ||
AY578142.1 | - | 6 | 948 | 73 | ||
EU556496.1 | - | 7 | 1125 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
