Assay Details
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044565.1 | NP_001038030.1 | 6 - 7 | 647 | 86 | |
GenBank mRNA | AK231666.1 | - | 6 - 7 | 666 | 86 | |
AK397319.1 | - | 6 - 7 | 712 | 86 | ||
DQ054778.1 | - | 6 - 7 | 664 | 86 | ||
DQ444460.1 | - | 6 - 7 | 647 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
