Assay Details
Gene Symbol: IFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213948.1 | NP_999113.1 | 1 - 2 | 142 | 90 | |
GenBank mRNA | AF493993.1 | - | 1 - 2 | 127 | 90 | |
AY188090.1 | - | 1 - 2 | 142 | 90 | ||
AY293733.1 | - | 1 - 2 | 127 | 90 | ||
DQ666352.1 | - | 1 - 2 | 118 | 90 | ||
DQ839398.1 | - | 1 - 2 | 142 | 90 | ||
DQ913893.2 | - | 1 - 2 | 185 | 90 | ||
EU118363.1 | - | 1 - 2 | 134 | 90 | ||
EU249804.1 | - | 1 - 2 | 118 | 90 | ||
GU433229.1 | - | 1 - 2 | 118 | 90 | ||
JF906510.1 | - | 1 - 2 | 118 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
