Assay Details
Gene Symbol: LDHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172363.2 | NP_001165834.1 | 6 - 7 | 812 | 82 | |
XM_013994501.2 | XP_013849955.2 | 1462 | 82 | |||
XM_021081651.1 | XP_020937310.1 | 895 | 82 | |||
XM_021081655.1 | XP_020937314.1 | 1203 | 82 | |||
XM_021081659.1 | XP_020937318.1 | 1024 | 82 | |||
GenBank mRNA | AK236433.1 | - | 6 - 7 | 850 | 82 | |
AK343612.1 | - | 6 - 7 | 803 | 82 | ||
AK391205.1 | - | 6 - 7 | 803 | 82 | ||
AK391657.1 | - | 6 - 7 | 804 | 82 | ||
AK395255.1 | - | 6 - 7 | 836 | 82 | ||
AK399432.1 | - | 6 - 7 | 804 | 82 | ||
AK399447.1 | - | 6 - 7 | 836 | 82 | ||
AK399466.1 | - | 6 - 7 | 803 | 82 | ||
AK399472.1 | - | 6 - 7 | 819 | 82 | ||
AK399544.1 | - | 6 - 7 | 803 | 82 | ||
AK399555.1 | - | 6 - 7 | 803 | 82 | ||
AK399560.1 | - | 8 - 9 | 1012 | 82 | ||
FJ865398.1 | - | 6 - 7 | 813 | 82 | ||
U07178.1 | - | 6 - 7 | 799 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
