Assay Details
Gene Symbol: C3H16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003354616.4 | XP_003354664.3 | 698 | 80 | ||
GenBank mRNA | AK390455.1 | - | 1 - 2 | 700 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
