Assay Details
Gene Symbol: KDELR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244970.1 | NP_001231899.1 | 2 - 3 | 368 | 102 | |
GenBank mRNA | AK236601.1 | - | 2 - 3 | 368 | 102 | |
AK397568.1 | - | 2 - 3 | 282 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
