Assay Details
Gene Symbol: NDUFAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243606.1 | NP_001230535.1 | 2 - 3 | 293 | 89 | |
GenBank mRNA | AK350073.1 | - | 2 - 3 | 279 | 89 | |
EW001058.2 | - | 2 - 3 | 300 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
