Assay Details
Gene Symbol: MANF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244655.1 | NP_001231584.1 | 1 - 2 | 125 | 97 | |
GenBank mRNA | AK233902.1 | - | 1 - 2 | 125 | 97 | |
AY609543.1 | - | 1 - 2 | 110 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
