Assay Details
Gene Symbol: MIR221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_035371.1 | 1 | 32 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | divergent protein kinase domain 2B | DIPK2B | CXHXorf36 | -348.1kb | Sense | Chr. X - 40179289 - 40223011 on NCBI Build Sscrofa11.1 |
Downstream | microRNA mir-222 | MIR222 | ssc-mir-222 | 671.0 | Sense | Chr. X - 40571808 - 40571887 on NCBI Build Sscrofa11.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
