Assay Details
Gene Symbol: TSPAN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021083873.1 | XP_020939532.1 | 142 | 122 | ||
XM_021083874.1 | XP_020939533.1 | 142 | 122 |

More Information
Set Membership: |
![]() ![]() |
