Assay Details
Gene Symbol: PTCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021098806.1 | XP_020954465.1 | 596 | 72 | ||
XM_021098807.1 | XP_020954466.1 | 739 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
