Assay Details
Gene Symbol: ZNF483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021085145.1 | XP_020940804.1 | 882 | 91 | ||
GenBank mRNA | AK233764.1 | - | 4 - 5 | 787 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
