Assay Details
Gene Symbol: SERHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021091433.1 | XP_020947092.1 | 414 | 118 | ||
XM_021091434.1 | XP_020947093.1 | 538 | 118 | |||
GenBank mRNA | AK234182.1 | - | 4 - 5 | 353 | 118 | |
AK389251.1 | - | 4 - 5 | 351 | 118 | ||
AK394188.1 | - | 4 - 5 | 353 | 118 | ||
AK398374.1 | - | 4 - 5 | 351 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
