Assay Details
Gene Symbol: RWDD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013992798.2 | XP_013848252.1 | 116 | 85 | ||
GenBank mRNA | AK230738.1 | - | 1 - 2 | 57 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013992798.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
