Assay Details
Gene Symbol: CDC42EP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131243.5 | XP_003131291.1 | 445 | 141 | ||
GenBank mRNA | AK236213.1 | - | 1 - 2 | 137 | 141 | |
AK394175.1 | - | 1 - 2 | 128 | 141 | ||
AK400730.1 | - | 1 - 2 | 155 | 141 | ||
AK400815.1 | - | 1 - 2 | 128 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003131243.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
