Assay Details
Gene Symbol: ZBTB32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013998339.2 | XP_013853793.1 | 1038 | 64 | ||
XM_021097178.1 | XP_020952837.1 | 100 | 64 | |||
GenBank mRNA | AK236945.1 | - | 3 - 4 | 270 | 64 | |
AK347589.1 | - | 2 - 3 | 1016 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
