Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003481554.4 | XP_003481602.1 | 1163 | 53 | ||
XM_013997533.2 | XP_013852987.1 | 911 | 53 | |||
GenBank mRNA | AK238002.1 | - | 2 - 3 | 886 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
