Assay Details
Gene Symbol: TNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013998200.2 | XP_013853654.1 | 1266 | 74 | ||
GenBank mRNA | AK343792.1 | - | 8 - 9 | 941 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
