Assay Details
Gene Symbol: TNFRSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068044.1 | XP_020923703.1 | 141 | 90 | ||
GenBank mRNA | AK350344.1 | - | 1 - 2 | 79 | 90 | |
AK397710.1 | - | 1 - 2 | 73 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021068044.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
