Assay Details
Gene Symbol: EPB41L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021075139.1 | XP_020930798.1 | 666 | 72 | ||
XM_021075140.1 | XP_020930799.1 | 576 | 72 | |||
XM_021075141.1 | XP_020930800.1 | 666 | 72 | |||
XM_021075142.1 | XP_020930801.1 | 433 | 72 | |||
XM_021075143.1 | XP_020930802.1 | 679 | 72 | |||
GenBank mRNA | AK232106.1 | - | 7 - 8 | 598 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
