Assay Details
Gene Symbol: COMMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005668159.3 | XP_005668216.1 | 529 | 95 | ||
GenBank mRNA | AK394424.1 | - | 6 - 7 | 533 | 95 | |
AY609968.1 | - | 6 - 7 | 514 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
