Assay Details
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021067109.1 | XP_020922768.1 | 4890 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021067109.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
