Assay Details
Gene Symbol: BNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003359404.4 | XP_003359452.3 | 600 | 82 | ||
GenBank mRNA | AK232439.1 | - | 4 - 5 | 529 | 82 | |
AK240169.1 | - | 4 - 5 | 82 | |||
AK349665.1 | - | 4 - 5 | 82 | |||
AK351253.1 | - | 4 - 5 | 82 | |||
AK398309.1 | - | 4 - 5 | 82 | |||
AK399168.1 | - | 4 - 5 | 556 | 82 | ||
AK400570.1 | - | 4 - 5 | 533 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
