Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021073458.1 | XP_020929117.1 | 1099 | 96 | ||
GenBank mRNA | AK232485.1 | - | 2 - 3 | 834 | 96 | |
AK393725.1 | - | 2 - 3 | 863 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
