Assay Details
Gene Symbol: MINDY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021094552.1 | XP_020950211.1 | 1919 | 119 | ||
GenBank mRNA | AK232627.1 | - | 8 - 9 | 1874 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
