Assay Details
Gene Symbol: BNIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977844.2 | XP_013833298.2 | 2844 | 64 | ||
XM_013977845.2 | XP_013833299.2 | 2135 | 64 | |||
XM_013977846.2 | XP_013833300.2 | 2723 | 64 | |||
XM_021098639.1 | XP_020954298.1 | 2802 | 64 | |||
XM_021098640.1 | XP_020954299.1 | 2015 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
