Assay Details
Gene Symbol: BNIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977844.2 | XP_013833298.2 | 2513 | 88 | ||
XM_013977845.2 | XP_013833299.2 | 1804 | 88 | |||
XM_013977846.2 | XP_013833300.2 | 2392 | 88 | |||
XM_021098639.1 | XP_020954298.1 | 2471 | 88 | |||
XM_021098640.1 | XP_020954299.1 | 1684 | 88 |
