Assay Details
Gene Symbol: RETREG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003131384.4 | XP_003131432.1 | 87 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003131384.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
