Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086504.1 | XP_020942163.1 | 1561 | 79 | ||
XM_021086505.1 | XP_020942164.1 | 1931 | 79 | |||
XM_021086506.1 | XP_020942165.1 | 1561 | 79 | |||
XM_021086507.1 | XP_020942166.1 | 1906 | 79 | |||
XM_021086508.1 | XP_020942167.1 | 1931 | 79 | |||
XM_021086509.1 | XP_020942168.1 | 1577 | 79 | |||
XM_021086510.1 | XP_020942169.1 | 1904 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
