Assay Details
Gene Symbol: ZNF598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013987817.2 | XP_013843271.2 | 1267 | 62 | ||
XM_013987818.2 | XP_013843272.2 | 1258 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
