Assay Details
Gene Symbol: MAD2L1BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013977970.2 | XP_013833424.1 | 265 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013977970.2 transcripts.
Set Membership: |
![]() ![]() |
