Assay Details
Gene Symbol: KBTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003129820.4 | XP_003129868.2 | 128 | 72 | ||
XM_003357271.4 | XP_003357319.1 | 119 | 72 | |||
XM_005667293.3 | XP_005667350.1 | 119 | 72 | |||
XM_021062727.1 | XP_020918386.1 | 119 | 72 | |||
XM_021062729.1 | XP_020918388.1 | 119 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003129820.4, XM_003357271.4, XM_005667293.3, XM_021062727.1, XM_021062729.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
