Assay Details
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244353.1 | NP_001231282.1 | 1 - 2 | 145 | 82 | |
GenBank mRNA | AK343994.1 | - | 1 - 2 | 145 | 82 | |
AK345051.1 | - | 1 - 2 | 146 | 82 | ||
AK391761.1 | - | 1 - 2 | 155 | 82 | ||
AK398730.1 | - | 1 - 2 | 167 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
