Assay Details
Gene Symbol: PSMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244353.1 | NP_001231282.1 | 3 - 4 | 329 | 92 | |
GenBank mRNA | AK343994.1 | - | 3 - 4 | 329 | 92 | |
AK391761.1 | - | 3 - 4 | 339 | 92 | ||
AK398730.1 | - | 3 - 4 | 351 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
