Assay Details
Gene Symbol: Myb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY615198.1 | - | 720 | 80 |
Gene Symbol: LOC100873106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JF951912.1 | - | 669 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
