Uvex by Honeywell Stealth Goggles
Uvex Stealth™
High-end styling for total comfort and unmatched protection from chemical splash and impact. Features a toric-shaped lens, soft elastomer body, quick-adjust headband and Uvextreme™ Anti-Fog lens coating.
- Neoprene headband
- Indirect vent
- Rx lens insert available
- Meets ANSI Z87.1-2010 and CSA Z94.3 standards
Uvex Stealth™ OTG
Over-the-Glass Goggle. Features the same high-performance protection against impact, chemical splash, and airborne particles as Uvex Stealth, and fits comfortably over most prescription eyewear.
- Neoprene headband
- Dura-streme™ dual anti-fog/anti-scratch lens coating
- Meets ANSI Z87.1-2010 and CSA Z94.3 standards
Uvex Stealth™ with HydroShield™ Lens Coating
One of the most popular, best-selling chemical splash goggles on the market now with revolutionary HydroShield anti-fog lens coating. Dual-action coating provides up to 60 times longer lasting fog-free vision than competitive anti-fog coated products. Meets ANSI Z87.1-2010 and CSA Z94.3 standards.