siRNA Details
Gene Symbol: BAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291428.1 | NP_001278357.1 | 4, 5 | 430 | ||
NM_001291429.1 | NP_001278358.1 | 3, 4 | 378 | |||
NM_001291430.1 | NP_001278359.1 | 2, 3 | 231 | |||
NM_001291431.1 | NP_001278360.1 | 2, 3 | 231 | |||
NM_004324.3 | NP_004315.1 | 4, 5 | 430 | |||
NM_138761.3 | NP_620116.1 | 4, 5 | 430 | |||
NM_138763.3 | NP_620118.1 | 3, 4 | 283 | |||
NM_138764.4 | NP_620119.2 | 4, 5 | 430 | |||
NR_027882.1 | 4, 5 | 430 | ||||
XM_017027077.1 | XP_016882566.1 | 4, 5 | 439 | |||
GenBank mRNA | AF007826.1 | - | 4, 5 | 361 | ||
AF247393.1 | - | 4, 5 | 361 | |||
AF250190.1 | - | 2, 3 | 162 | |||
AJ417988.1 | - | 3, 4 | 340 | |||
AJ586909.1 | - | 2, 3 | 149 | |||
AJ586910.1 | - | 3, 4 | 296 | |||
AK291076.1 | - | 4, 5 | 410 | |||
BC014175.2 | - | 4, 5 | 413 | 4578562 | ||
BE396495.1 | - | 4 | 430 | |||
BM706954.1 | - | 2, 3 | 243 | |||
CN281003.1 | - | 4, 5 | 383 | |||
HQ889830.1 | - | 2, 3 | 162 | |||
JX524562.1 | - | 3, 4 | 310 | |||
KF830846.1 | - | 3, 4 | 310 | |||
KF830847.1 | - | 3, 4 | 310 | |||
KF830848.1 | - | 3, 4 | 310 | |||
L22473.1 | - | 4, 5 | 361 | |||
L22474.1 | - | 4, 5 | 361 | |||
T29044.1 | - | 1, 2 | 98 | |||
U19599.1 | - | 3, 4 | 214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
