Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ 80nm Colloidal Gold Nanoparticles (15OD)
Shop All Novus Biologicals Products

Click to view available options
Quantity:
100 mL
Description
80 nm colloidal gold is an aqueous suspension of spherical metallic nanoparticles manufactured to the highest standard to ensure:
- Uniform spherical shape
- Narrow size distribution: 80 nm ± 8nm, <10% CV
- Ultra high quality
- Batch-to-batch consistency
- High efficiency antibody binding for maximum sensitivity
- Available at high concentrations to simplify assay development
Specifications
Specifications
Quantity | 10 mL |
Content And Storage | 4°C Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction