Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ABCF3 Overexpression Lysate (Native)
SDP

Catalog No. NBL107185 Shop All Novus Biologicals Products
Click to view available options
:
The ABCF3 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.

Specifications

For Use With (Application)

Western Blot

Cell Line

The lysate was created in HEK293T cells, using plasmid ID RC2143 and based on accession number NM_18358. The protein contains a C-terminal DDK tag.

Quantity

0.1 mg

Storage Buffer

RIPA buffer

Purity

Protein

Test Specificity

Homo sapiens ATP-binding cassette, sub-family F (GCN20), member 3 (ABCF3), mRNA.

Content And Storage

-80°C. Avoid freeze-thaw cycles.

Protein State

Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.