Learn More
Novus Biologicals™ ABI1 Overexpression Lysate (Native)
Shop All Novus Biologicals Products

Specifications
For Use With (Application)
Western Blot
Cell Line
The lysate was created in HEK293T cells, using plasmid ID RC21264 and based on accession number NM_112752. The protein contains a C-terminal DDK tag.
Quantity
0.1 mg
Storage Buffer
RIPA buffer
Purity
Protein
Test Specificity
Homo sapiens abl-interactor 1 (ABI1), transcript variant 4, mRNA.
Content And Storage
-80°C. Avoid freeze-thaw cycles.
Protein State
Native
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.