Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ AbPure Antibody Concentration and Clean-Up Antibody Purification Kit
SDP

Catalog No. 2620010NB Shop All Novus Biologicals Products
Click to view available options
:
Sufficient for 3 columns

Allows removal of contaminants that interfere in conjugation reactions of InnovaCoat Gold

Kit Components
  • 3 spin cartridge/collecting tube assemblies
  • 1 bottle of AbPure Conjugation Buffer

Specifications

Quantity 3 Tests
Content And Storage 4°C
Kit Contents 3 spin cartridge/collecting tube assemblies, 1 bottle of AbPure Conjugation Buffer
Sufficient For 3 columns
Product Type AbPure Antibody Concentration and Clean-Up Antibody Purification Kit

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.