Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenosine A1R Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 6 publications

Supplier:  Novus Biologicals NB3005490.025ML

 View more versions of this product

Catalog No. NB414035


Explore all special offers
Add to Cart

Description

Description

Adenosine A1R Polyclonal specifically detects Adenosine A1R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Adenosine A1R
Polyclonal
Unconjugated
PBS and 1 mg/ml BSA with 0.05% Sodium Azide
adenosine A1 receptor, ADORA 1, RDC7adenosine receptor A1
Rabbit
37 kDa
0.025 mL
Primary
Detects Adenosine Receptor A1. This does not detect other AR subtypes.
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1:500, Flow Cytometry 3 - 5 μg/ 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen
P25099
ADORA1
Synthetic peptide corresponding to residues C(309) Q P K P P I D E D L P E E K A E D(326) of rat Adenosine Receptor A1.
Affinity Purified
RUO
134
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only