Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine A1R Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
$196.10
Specifications
Antigen | Adenosine A1R |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500, Flow Cytometry 3 - 5 μg/ 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen |
Applications | Western Blot |
Classification | Polyclonal |
Description
Adenosine A1R Polyclonal specifically detects Adenosine A1R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Adenosine A1R | |
Western Blot 1:500, Flow Cytometry 3 - 5 μg/ 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Immunohistochemistry-Frozen | |
Polyclonal | |
Rabbit | |
Human | |
P25099 | |
134 | |
Synthetic peptide corresponding to residues C(309) Q P K P P I D E D L P E E K A E D(326) of rat Adenosine Receptor A1. | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. | |
37 kDa |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 1 mg/ml BSA with 0.05% Sodium Azide | |
adenosine A1 receptor, ADORA 1, RDC7adenosine receptor A1 | |
ADORA1 | |
IgG | |
Affinity Purified | |
Detects Adenosine Receptor A1. This does not detect other AR subtypes. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title