Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Adenosine A2aR Recombinant Protein Antigen
SDP

Catalog No. NBP62698PEP Shop All Novus Biologicals Products
Click to view available options
:

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ADORA2A. The Adenosine A2aR Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez)

135

Species

Human

Purity

>80% by SDS-PAGE and Coomassie blue staining

Common Name

Adenosine A2aR

Content And Storage

Store at -20°C. Avoid freeze-thaw cycles.

Formulation

PBS and 1M Urea, pH 7.4.

For Use With (Application)

Antibody Competition

Gene Symbol

ADORA2A

Molecular Weight (g/mol)

23 kDa

Quantity

100 μl

Regulatory Status

RUO

Source

E. coli

Specific Reactivity

This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54484. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.