Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine Deaminase/ADA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30036450UL
Description
Adenosine Deaminase/ADA Polyclonal antibody specifically detects Adenosine Deaminase/ADA in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
Adenosine Deaminase/ADA | |
Polyclonal | |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
Produced in rabbits immunized with purified, recombinant Human Adenosine Deaminase/ADA (Uniprot#: P00813; Met1-Leu363). | |
50 μg | |
Cancer, Endocrinology | |
100 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction