Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenosine Deaminase/ADA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Adenosine Deaminase/ADA |
---|---|
Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate |
Applications | Western Blot, ELISA, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Adenosine Deaminase/ADA Polyclonal antibody specifically detects Adenosine Deaminase/ADA in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
Adenosine Deaminase/ADA | |
Western Blot, ELISA, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology | |
0.2 um filtered solution in PBS | |
100 | |
IgG | |
Antigen and protein A Affinity-purified |
Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ADA1, adenine deaminase, Adenosine aminohydrolase, adenosine deaminase, EC 3.5.4.4 | |
Produced in rabbits immunized with purified, recombinant Human Adenosine Deaminase/ADA (Uniprot#: P00813; Met1-Leu363). | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title