Learn More
AIMS™ Traditional Triangle Shaped Plastic Disposable Restraints
Humane, safe and disposable rodent restraint system
Supplier:  AIMS™ 1803B
 View more versions of this product
Catalog No. 14-370-109
Explore all special offers
Add to Cart
Includes: Instructions and twist ties
Description
Description
This rodent restraint system is available in a variety of sizes and colors. The cone-shaped bags provide easier animal capture and increased animal comfort.
  • Precut sizes for quick use
  • Easy-Flex-PETM provides effortless wrapping and added animal comfort
  • Body-Contour Cone-Shape provides easier restraint, exceptional comfort, and reduced animal stress
  • Black and rose color bags reduce natural light and aids in reducing animal agitation and stress
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Specifications
Specifications
Black
Restraint
Plastic
Large
Tattooing, Tail Vein Injections, Oral Dosing, Decapitation and General Restraint
Yes
Triangle
≤450 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.