Learn More
Cyclin B Mouse, Unlabeled, Clone: 18, BD
Mouse Monoclonal Antibody
$286.00 - $526.00
Specifications
Antigen | Cyclin B |
---|---|
Clone | 18 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The B-type cyclins serve as subunits of the major mitotic protein kinase in many different organisms. In mammals, Cyclin B is a subunit for the cdc2 (Cdk1) protein kinase and is synthesized during late S and G2 phases. Cyclin B associates with inactive cdc2 and facilitates phosphorylation of cdc2 at Thr-14 and Tyr-15. This phosphorylation maintains the inactive state until the end of G2. The inactive Cyclin B-cdc2 complex continues to accumulate in the cytoplasm until the CDC25 phosphatase dephosphorylates Thr-14 and Tyr-15, rendering the complex active at the G2/M boundary. This mitotic kinase complex remains active until the metaphase/anaphase transition when Cyclin B becomes degraded. This degradation process appears to be ubiquitin-dependent and is necessary for the cell to exit mitosis. Also, the conserved domain in B-type cyclins, known as the P box, is required for CDC25 activation.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Cyclin B | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
18 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.